Order Entry
United States
Orders LinkContactUsLinkComponent
Thymolphthalein ACS

Thymolphthalein ACS

Plastic bottle, 100 g
Catalog # CH01270-100G
New Product
Supplier #:  01270-100G
Thymolphthalein ACS
Catalog # CH01270-100G
New Product
CAS Number:  125-20-2
Thymolphthalein ACS
Thymolphthalein ACS

Plastic bottle, 100 g

CAS Number:  125-20-2
Price
$132.71
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Thymolphthalein is a versatile chemical compound widely recognized for its application as a pH indicator in various laboratory settings. This compound transitions from colorless to blue as the pH of a solution shifts from acidic to alkaline, making it an essential tool for researchers and professionals in fields such as chemistry, biology, and environmental science. Its unique properties allow for precise monitoring of pH levels, which is crucial in processes like titrations and the formulation of pharmaceuticals.

In addition to its role as a pH indicator, Thymolphthalein is also utilized in the synthesis of dyes and pigments, enhancing its relevance in the textile and paint industries. Its stability and effectiveness in a range of conditions make it a preferred choice for professionals seeking reliable performance. With its dual functionality in both analytical and industrial applications, Thymolphthalein stands out as a valuable compound for those looking to optimize their processes and achieve accurate results.

Store at room temperature.

Synonyms:  3,3-Bis(4-hydroxy-5-isopropyl-o-tolyl)phthalide, 3,3-Bis(4-hydroxy-5-isopropyl-2-methylphenyl)isobenzofuran-1(3H)-one
  • CAS Number:125-20-2
  • Formula:C₂₈H₃₀O₄
  • MDL Number:MFCD00005909
  • Molecular Weight:430.5 g/mol
  • Storage Temperature:Room Temperature
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.