Order Entry
United States
Orders LinkContactUsLinkComponent
Ethanol ≥95%

Ethanol ≥95%

500 ml
Catalog # 77974-111
New Product
Supplier #:  051-00476
Ethanol ≥95%
Catalog # 77974-111
New Product
CAS Number:  64-17-5
Ethanol ≥95%
Ethanol ≥95%

500 ml

CAS Number:  64-17-5
Price
$31.20
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Ethanol (95) is a high-purity, clear colorless liquid used primarily in research applications.

Caution: For research use only (RUO).

Synonyms:  Ethyl alcohol
  • CAS Number:64-17-5
  • Formula:C₂H₅OH
  • Molecular Weight:46.07 g/mol
  • Storage Temperature:Ambient
Specifications

Specification Test Results

  • Assay:
    94.8 - 95.8 vol% by density

Specifications

  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.