(±)-Epichlorohydrin ≥99% (by GC)
Plastic bottle, 1 kg
Catalog # CH24484-1KG
New Product
Supplier: Chem-Impex International
Supplier #: 24484-1KG
(±)-Epichlorohydrin ≥99% (by GC)
Catalog # CH24484-1KG
Supplier: Chem-Impex International
New Product
CAS Number: 106-89-8
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Shipping temperature: Store at 0 to 8 °C
Synonyms: (±)-1-Chloro-2,3-epoxypropane, Epichlorohydrin, (±)-3-Chloro-1,2-epoxypropane, 1-Chloro-2,3-epoxypropane, Epichlorhydrin, (±)-Glycerol epichlorhydrin, 3-Chloro-1,2-epoxypropane
- CAS Number:106-89-8
- Formula:C₃H₅ClO
- MDL Number:MFCD00005132
- Molecular Weight:92.52 g/mol
- Storage Temperature:Refrigerator
- UN:2023
Specifications
Specifications
- Pack type:Plastic bottle
- Size:1 kg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



