Order Entry
United States
Orders LinkContactUsLinkComponent
D(+)-Mannose ≥99% (by HPLC)

D(+)-Mannose ≥99% (by HPLC)

Plastic bottle, 250 g
Catalog # CH01458-250G
New Product
Supplier #:  01458-250G
D(+)-Mannose ≥99% (by HPLC)
Catalog # CH01458-250G
New Product
CAS Number:  3458-28-4
D(+)-Mannose ≥99% (by HPLC)
D(+)-Mannose ≥99% (by HPLC)

Plastic bottle, 250 g

CAS Number:  3458-28-4
Price
$56.40
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

D(+)-Mannose is a naturally occurring sugar that plays a vital role in various biological processes. This simple monosaccharide is recognized for its potential applications in the pharmaceutical and nutraceutical industries, particularly for its ability to support urinary tract health. D(+)-Mannose works by preventing the adhesion of certain bacteria to the urinary tract walls, thereby reducing the risk of infections. Its unique structure allows for easy absorption and utilization by the body, making it a preferred choice for dietary supplements aimed at promoting urinary health.

In addition to its health benefits, D(+)-Mannose is also utilized in research settings, particularly in studies related to carbohydrate metabolism and cell signaling. Its versatility extends to food and beverage formulations, where it can serve as a natural sweetener with low caloric content. With its growing recognition in both health and research sectors, D(+)-Mannose stands out as a valuable compound for professionals seeking effective solutions for urinary health and metabolic studies.

Shipping temperature: Store at RT

Synonyms:  (2S,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal
  • CAS Number:3458-28-4
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00064122
  • Molecular Weight:180.16 g/mol
  • Storage Temperature:Room Temperature
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    250 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.