Order Entry
United States
Orders LinkContactUsLinkComponent
Potassium hydroxide 85-100.5% (total base), pellets FCC

Potassium hydroxide 85-100.5% (total base), pellets FCC

New Product
Potassium hydroxide 85-100.5% (total base), pellets FCC
New Product
Certificates
Potassium hydroxide 85-100.5% (total base), pellets FCC
Potassium hydroxide 85-100.5% (total base), pellets FCC

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Potassium hydroxide, commonly known as caustic potash, is a versatile and essential chemical compound widely used across various industries. This strong alkaline substance is available in pellet form, making it easy to handle and store. Potassium hydroxide is primarily utilized in the production of fertilizers, where it serves as a key ingredient in nutrient solutions for crops, enhancing their growth and yield. Additionally, it plays a crucial role in the manufacturing of soaps and detergents, acting as a saponifying agent that helps in the conversion of fats and oils into usable products.

In the laboratory, potassium hydroxide is frequently employed in titrations and as a reagent in various chemical syntheses, demonstrating its importance in research and analytical applications. Its ability to absorb moisture and carbon dioxide makes it valuable in moisture control applications as well. With its high solubility in water and strong alkaline properties, potassium hydroxide stands out as a reliable choice for professionals seeking effective solutions in agriculture, cleaning products, and chemical manufacturing.

Shipping temperature: Store at RT

Synonyms:  Potash caustic, Potassium hydrate, Caustic potash
  • CAS Number:1310-58-3
  • Formula:KOH
  • MDL Number:MFCD00003553
  • Molecular Weight:56.1 g/mol
  • Storage Temperature:Room Temperature
  • UN:1813
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Plastic bottle
    1 kg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $45.00
    $45.00
    Each(1/Itm/Bx)
       $45.00

    Supplier Number: 00252-1KG
    Specifications:
    New Product
    New Product
  • Plastic bottle
    5 kg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $180.01
    $180.01
    Each(1/Itm/Bx)
       $180.01

    Supplier Number: 00252-5KG
    Specifications:
    New Product
    New Product
  • Plastic bottle
    100 kg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $2,150.00
    $2,150.00
    Each(1/Itm/Bx)
       $2,150.00

    Supplier Number: 00252-100KG
    Specifications:
    New Product
    New Product
  • Plastic bottle
    25 kg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $650.00
    $650.00
    Each(1/Itm/Bx)
       $650.00

    Supplier Number: 00252-25KG
    Specifications:
    New Product
    New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.