Orders LinkContactUsLinkComponent
PathCam Mobile Imaging Stations
PathCam Mobile Imaging Stations
New Product
Supplier:   MOPEC
PathCam Mobile Imaging Stations
Supplier:  MOPEC
New Product
Certificates
PathCam Mobile Imaging Stations
PathCam Mobile Imaging Stations
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Mobile imaging stations offer a fully integrated, high-definition imaging solution designed for flexibility and precision in pathology and autopsy environments.
  • Mobile imaging station for monitor, keyboard, and PathCam systems
  • Compatible with PathCam HD and HR cameras
  • AX047 PathCam foot pedal command
  • 4 heavy-duty casters
  • Articulating monitor arm and mount
  • Articulating arm with platform for keyboard and mouse
  • Articulating camera arm with 45” reach
  • Base includes four access doors
Built with a durable stainless steel base, articulating arms, and silent-grade casters, these stations support the PathCam HD or HR for seamless image capture, live streaming, and documentation. The ergonomic, mobile design with advanced connectivity and cable management enhances workflow efficiency and remote collaboration.
Stations include column and base, articulating camera arm (18 - 47.5" adjustable reach from column to camera), keyboard and monitor arm, On/Off power switch for camera, removable foot pedal shelf, pull handle for transport, Wi-Fi antenna, tension knob for precise camera pivot, power cable port, Ethernet port, USB ports, and cable management system.
Manufactured from a combination of powder-coated cast aluminum grade 6061 and 304 stainless steel with a polished no. four finish.
Product Family Options
Product Information
  • Availability
    Price
  • 78141-958
    PathCam HD mobile imaging station
    PathCam HD mobile imaging station
    Log in for availability
    Log in for availability
    Each
    Each - $13,264.52
    $13,264.52
    Each
       $13,264.52
    Supplier Number: AX260HD
    Specifications:
    New Product
    New Product
  • 78141-959
    PathCam HR mobile imaging station
    PathCam HR mobile imaging station
    Log in for availability
    Log in for availability
    Each
    Each - $13,714.23
    $13,714.23
    Each
       $13,714.23
    Supplier Number: AX260HR
    Specifications:
    New Product