Orders LinkContactUsLinkComponent
PathCam Wall Mount Systems
PathCam Wall Mount Systems
New Product
Supplier:   MOPEC
PathCam Wall Mount Systems
Supplier:  MOPEC
New Product
Certificates
PathCam Wall Mount Systems
PathCam Wall Mount Systems
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The PathCam wall mount system enhances pathology workflows by integrating an articulating keyboard arm, monitor arm, and camera arm for ergonomic, space-saving digital documentation.
  • Compatible with PathCam HD and HR cameras
  • Equipped with several pivot points to articulate up and down, and side to side
  • Can pivot at the wall, swivel, and tilt to optimize camera-viewing angles
  • Can be pulled out when in use, then pushed back when not in use
Designed for use at grossing stations, the system improves efficiency, accuracy, and collaboration in clinical settings through seamless integration of digital systems. The articulating camera arm has a minimum reach of 20", and a maximum reach of 62" (wall surface to camera-mounting mid-point).
Systems include (1) 42” wall mount track, (1) 10” wall mount track, (1) articulating camera arm with 60” reach, (1) vertical CPU holder for 10” wall track, and mounting hardware for wall tracks.
Manufactured from a combination of powder-coated cast aluminum grade 6061 and 304 stainless steel with a polished no.four finish.
Product Family Options
Product Information
  • Availability
    Price
  • 78141-956
    PathCam HD wall mount system
    PathCam HD wall mount system
    Log in for availability
    Log in for availability
    Each
    Each - $13,263.14
    $13,263.14
    Each
       $13,263.14
    Supplier Number: AX250HD
    Specifications:
    New Product
    New Product
  • 78141-957
    PathCam HR wall mount system
    PathCam HR wall mount system
    Log in for availability
    Log in for availability
    Each
    Each - $13,714.23
    $13,714.23
    Each
       $13,714.23
    Supplier Number: AX250HR
    Specifications:
    New Product