Orders LinkContactUsLinkComponent
Gradient Makers
Gradient Makers
Marketsource
Supplier:   HOEFER INC MS
Gradient Makers
Supplier:  HOEFER INC MS
Marketsource
Certificates
Gradient Makers
5 Options Available Below
Product Details & Documents
Ideal for generating gradients of polyacrylamide, sucrose and cesiium chloride.
  • Milled of heavy acrylic plastic with high quality, leak-free Teflon® valves
  • Flat base provides stability on magnetic stir
  • Comes with a support rod for attaching to a ring stand
  • Takes Luer or 4 mm int.Ø tubing
  • With included adapters, can also take 2 mm int.Ø or 22-gauge tubing
The 500 ml SG500 Gradient Maker is made of two cylindrical acrylic chambers joined and mounted on a flat acrylic base. An easy-to-use push-pull valve opens and closes the passage between the reservoir and mixing chambers. Suitable for forming polyacrylamide gradients or buffer gradients used with medium to small chromatography columns. Ideal for use with the SE615 Multiple Gel Caster.
Ordering information: SG15, SG30, SG50 and SG100 include Gradient Maker with Teflon Valves; Adapter Barbed Fitting; Luer Outlet Fitting; 22-gauge Needle; Support Rod. SG500 includes Gradient Maker with Push-Pull Valve; Hose Clamps (4 pieces).
Product Family Options
Product Information
  • Availability
    Price
  • MSPP-SG100
    Gradient maker, 100 ml total volume
    Gradient maker, 100 ml total volume
    Log in for availability
    Log in for availability
    Each
    Each - $650.00
    $650.00
    Each
       $650.00
    Minimum order quantity is 1
    Supplier Number: SG100
    Specifications:
    Marketsource
    Marketsource
  • MSPP-SG15
    Gradient maker, 15 ml total volume
    Gradient maker, 15 ml total volume
    Log in for availability
    Log in for availability
    Each
    Each - $540.00
    $540.00
    Each
       $540.00
    Minimum order quantity is 1
    Supplier Number: SG15
    Specifications:
    Marketsource
    Marketsource
  • MSPP-SG30
    Gradient maker, 30 ml total volume
    Gradient maker, 30 ml total volume
    Log in for availability
    Log in for availability
    Each
    Each - $550.00
    $550.00
    Each
       $550.00
    Minimum order quantity is 1
    Supplier Number: SG30
    Specifications:
    Marketsource
    Marketsource
  • MSPP-SG50
    Gradient maker, 50 ml total volume
    Gradient maker, 50 ml total volume
    Log in for availability
    Log in for availability
    Each
    Each - $610.00
    $610.00
    Each
       $610.00
    Minimum order quantity is 1
    Supplier Number: SG50
    Specifications:
    Marketsource
    Marketsource
  • MSPP-SG500
    Gradient maker, 500 ml total volume
    Gradient maker, 500 ml total volume
    Log in for availability
    Log in for availability
    Each
    Each - $740.00
    $740.00
    Each
       $740.00
    Minimum order quantity is 1
    Supplier Number: SG500
    Specifications:
    Marketsource
    Marketsource