Order Entry
United States
Orders LinkContactUsLinkComponent
Copper(I) bromide ≥98%

Copper(I) bromide ≥98%

Bottle, 100 g
Catalog # ALC434113-100G
Irritant
Corrosive
Environment
New Product
Supplier:   Aladdin Scientific
Supplier #:  C434113-100G
Copper(I) bromide ≥98%
Catalog # ALC434113-100G
Supplier:  Aladdin Scientific
New Product
CAS Number:  7787-70-4
Copper(I) bromide ≥98%

Bottle, 100 g

CAS Number:  7787-70-4
Price
$17.90
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Copper (I) bromide is a copper (I) halide. Copper(I) bromide along with N -( n -octyl)-2-pyridylmethanimine participates in the living-radical polymerization of methyl methacrylate. It reacts with various heterocyclic thiones (L) in the presence of triphenylphosphine, to afford the mononuclear complexes of the general formula [Cu(L)(PPh 3 ) 2 Br]. Application: Copper (I) bromide may be used in the synthesis of the following: enantioenriched, functionalized and protected propargylamines polyvinylchloride bearing benzoxazine side groups polybutadiene having benzoxazine side-groups (PB-benzoxazine).

  • CAS Number:7787-70-4
  • Formula:BrCu
  • Molecular Weight:143.45 g/mol
  • Storage Temperature:Ambient
  • EINECS:232-131-6
Specifications

Specifications

  • Pack type:
    Bottle
  • Size:
    100 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.