Order Entry
United States
Orders LinkContactUsLinkComponent
α-Cyclodextrin ≥98% cell culture reagent

α-Cyclodextrin ≥98% cell culture reagent

Supplier:   Aladdin Scientific
α-Cyclodextrin ≥98% cell culture reagent
Supplier:  Aladdin Scientific
Certificates
α-Cyclodextrin ≥98% cell culture reagent

4 Options Available Below

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:10016-20-3
  • Formula:C₃₆H₆₀O₃₀
  • MDL Number:MFCD00078207
  • Molecular Weight:972.84 g/mol
  • Storage Temperature:Ambient
  • EINECS:233-007-4
Product Family Options

Product Information

  • Pack type
    Size
    Availability
    Price
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $142.00
    $142.00
    Each(1/Itm/Bx)
       $142.00

    Supplier Number: C106778-100G
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $9.90
    $9.90
    Each(1/Itm/Bx)
       $9.90

    Supplier Number: C106778-1G
  • Bottle
    25 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $39.90
    $39.90
    Each(1/Itm/Bx)
       $39.90

    Supplier Number: C106778-25G
  • Bottle
    5 g
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $25.90
    $25.90
    Each(1/Itm/Bx)
       $25.90

    Supplier Number: C106778-5G

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.