Hypericin ≥97% PKC inhibitor
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Selective PKC inhibitor (IC₅₀ = 3.3 μM). Antiviral agent. Potent tyrosine kinase inhibitor (IC₅₀ = 7.5 μM). Potent CKII inhibitor (IC₅₀ = 6 nM). Inactivates enveloped viruses including HIV. Promotes VEGF and TGF-β production. Shows photosensitizing and antidepressant effects in vivo. .
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- Molecular Weight:504.4 g/mol
Specifications
Specifications
- Size:1 mg