Order Entry
United States
Orders LinkContactUsLinkComponent
Lenalidomide ≥98% Thalidomide analog TNF alpha inhibitor

Lenalidomide ≥98% Thalidomide analog TNF alpha inhibitor

New Product
Supplier:   Abcam
Lenalidomide ≥98% Thalidomide analog TNF alpha inhibitor
Supplier:  Abcam
New Product
Certificates
Lenalidomide ≥98% Thalidomide analog TNF alpha inhibitor

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Thalidomide analog and TNF-α inhibitor. Potent antitumor and anti-inflammatory effects. Modulates and potentiates host immune responses against multiple myeloma. Active in vitro.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  3-(4-Amino-1-oxo-1,3-dihydro-2H-isoindol-2-yl)piperidine-2,6-dione, (R,S)-Lenalidomide, 3-(4-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione
  • CAS Number:191732-72-6
  • Molecular Weight:259.26 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% LENALIDOMIDE
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 100 mg
    Log in for availability
    Log in for availability
    Each (100mg)
    Each (100mg) - $311.00
    $311.00
    Each (100mg)
       $311.00
    Minimum order quantity is 1

    Supplier Number: AB142129-100MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
  • 25 mg
    Log in for availability
    Log in for availability
    Each (25mg)
    Each (25mg) - $122.00
    $122.00
    Each (25mg)
       $122.00
    Minimum order quantity is 1

    Supplier Number: AB142129-25MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product