Orders LinkContactUsLinkComponent
SD-169 ≥99% ATP-competitive p38alpha MAPK inhibitor
New Product
 :   Abcam
SD-169 ≥99% ATP-competitive p38alpha MAPK inhibitor
 :  Abcam
New Product
SD-169 ≥99% ATP-competitive p38alpha MAPK inhibitor
Potent, selective and ATP-competitive p38α MAPK inhibitor (IC₅₀ values are 3.2 and 122 nM for α and β, respectively). Orally active. Promotes axonal regeneration in vivo.
 : All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
 :  p38 MAP Kinase Inhibitor VII, 1H-indole-5-carboxamide, Indole-5-carboxamide
  •  :1670-87-7
  •  :160.17 g/mol
  • Constituents::
    100% 1H-Indole-5-carboxamide
Product Family Options
  • Size
  • 10 mg
    Each (10mg)
    Each (10mg) - $143.00
    $143.00
    Each (10mg)
       $143.00
     : AB120836-10MG
    New Product
    New Product
  • 50 mg
    Each (50mg)
    Each (50mg) - $564.00
    $564.00
    Each (50mg)
       $564.00
     : AB120836-50MG