Order Entry
United States
Orders LinkContactUsLinkComponent
α-Cyclodextrin non-reducing cyclic saccharide

α-Cyclodextrin non-reducing cyclic saccharide

New Product
Supplier:   Abcam
α-Cyclodextrin non-reducing cyclic saccharide
Supplier:  Abcam
New Product
Certificates
α-Cyclodextrin non-reducing cyclic saccharide

2 Options Available Below

Product Details & Documents

Non-reducing cyclic saccharide consisting of six α-1,4-linked D-glucopyranosyl units produced by the action of cyclodextrin glucosyltransferase (CGTase, EC 2.4.1.19) on hydrolyzed starch. Useful for the solubilization of nonpolar macromolecules such as fatty acids, lipids, and cholesterol for use in cell culture applications. Cyclodextrins are low molecular weight organic molecules which form water-soluble complexes with various compounds.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

  • CAS Number:10016-20-3
  • Molecular Weight:972.86 g/mol
Specifications for Product Family

Specification Test Results

  • Constituents::
    100% a-Cyclodextrin hydrate
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 25 g
    Log in for availability
    Log in for availability
    Each (25g)
    Each (25g) - $269.00
    $269.00
    Each (25g)
       $269.00
    Minimum order quantity is 1

    Supplier Number: AB146272-25G
    Specifications:
    New Product
    New Product
  • 5 g
    Log in for availability
    Log in for availability
    Each (5g)
    Each (5g) - $73.80
    $73.80
    Each (5g)
       $73.80
    Minimum order quantity is 1

    Supplier Number: AB146272-5G
    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.