D-(-)-Luciferin ≥98% chemiluminescent luciferase substrate
About this item
Cell-permeable chemiluminescent luciferase substrate. Emits lights upon oxidative decarboxylation in the presence of ATP. Provides a bioluminescent signal for cellular ATP levels in vivo. Water soluble analogs are D-Luciferin potassium salt and D-Luciferin sodium salt.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:2591-17-5
- Molecular Weight:280.3 g/mol
3 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Cell-permeable chemiluminescent luciferase substrate. Emits lights upon oxidative decarboxylation in the presence of ATP. Provides a bioluminescent signal for cellular ATP levels in vivo. Water soluble analogs are D-Luciferin potassium salt and D-Luciferin sodium salt.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
- CAS Number:2591-17-5
- Molecular Weight:280.3 g/mol
Specifications for Product Family
Specification Test Results
- Constituents::99.53% (S)-4,5-dihydro-2-(6-hydroxybenzothiazol-2-yl)thiazole-4-carboxylic acid
Product Family Options
Product Information
- SizeAvailabilityPrice
- Supplier Number: AB143654-10MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New ProductNew Product
- Supplier Number: AB143654-50MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New ProductNew Product
- Supplier Number: AB143654-5MGItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:New ProductNew Product



