Order Entry
United States
Orders LinkContactUsLinkComponent
Dexamethasone ≥99% anti-inflammatory glucocorticoid

Dexamethasone ≥99% anti-inflammatory glucocorticoid

100 mg
Catalog # ABCA_AB120743-100M
New Product
Supplier:   Abcam
Supplier #:  AB120743-100MG
Dexamethasone ≥99% anti-inflammatory glucocorticoid
Catalog # ABCA_AB120743-100M
Supplier:  Abcam
New Product
CAS Number:  50-02-2
Dexamethasone ≥99% anti-inflammatory glucocorticoid

100 mg

CAS Number:  50-02-2
Price
$89.60
Log in for availability
Each (100mg)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Anti-inflammatory glucocorticoid (IC₅₀ =38 nM and Ki =6.7 nM for the glucocorticoid receptor). Displays 7-fold higher affinity for glucocorticoid receptors than hydrocortisone. Long-term effects following oral administration. Used in the generation of liver organoids.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Dexamethasonum, 9-Fluor-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadien-3,20-dion
  • CAS Number:50-02-2
  • Molecular Weight:392.5 g/mol
Specifications

Specification Test Results

  • Constituents::
    100% Dexamethasone

Specifications

  • Size:
    100 mg

Specifications