Order Entry
United States
Orders LinkContactUsLinkComponent
Cycloheximide ≥98% Protein synthesis inhibitor

Cycloheximide ≥98% Protein synthesis inhibitor

New Product
Supplier:   Abcam
Cycloheximide ≥98% Protein synthesis inhibitor
Supplier:  Abcam
New Product
Certificates
Cycloheximide ≥98% Protein synthesis inhibitor

1 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Protein synthesis inhibitor. Antifungal antibiotic. Achieve your results faster with highly validated, pure and trusted compounds.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Synonyms:  Naramycin A, Actidione, 3-[2-(3,5-Dimethyl-2-oxocyclohexyl)-2-hydroxyethyl]glutarimide,
  • Molecular Weight:281.35 g/mol
Product Family Options

Product Information

  • Size
    Availability
    Price
  • 1 g
    Log in for availability
    Log in for availability
    Each (1g)
    Each (1g) - $116.00
    $116.00
    Each (1g)
       $116.00
    Minimum order quantity is 1

    Supplier Number: AB120093-1G
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
  • 100 mg
    Discontinued
    Supplier Number: AB120093-100MG
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.