Order Entry
United States
Orders LinkContactUsLinkComponent
Milwaukee® Digital Sugar Refractometer (Brix)

Milwaukee® Digital Sugar Refractometer (Brix)

Digital sugar refractometer (Brix), 0 - 85% Brix, ±0.2% Brix, ±0.3 °C, ±0.5 °F
Catalog # 470359-612
New Product
Supplier #:  MA871
Milwaukee® Digital Sugar Refractometer (Brix)
Catalog # 470359-612
New Product
Milwaukee® Digital Sugar Refractometer (Brix)
Milwaukee® Digital Sugar Refractometer (Brix)

Digital sugar refractometer (Brix), 0 - 85% Brix, ±0.2% Brix, ±0.3 °C, ±0.5 °F

Price
$199.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Ideal for student laboratories with minimal mess – only 2 to 3 drops needed and results appear instantly on a digital display.

  • This durable refractometer is easy to use and requires minimal maintenance Fast, accurate results with digital dual level LCD readout Automatically temperature compensated Simple one-point calibration using distilled or deionized water

Want accurate, reliable and repeatable results? This Milwaukee Refractometer makes learning hands on simple with fast, accurate readings with just a few drops – perfect for food science, chemistry, and beverage experiments in the classroom or laboratory.

Ordering information: Batteries included. Does NOT include distilled or deionized water for calibration.

Specifications
  • Description:
    Digital sugar refractometer (Brix)
  • Accuracy:
    ±0.2% Brix, ±0.3 °C, ±0.5 °F
  • Range:
    0 - 85% Brix
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.