Orders LinkContactUsLinkComponent
Milwaukee® pH/ORP Cleaning Solution
pH/ORP cleaning solution (230 ml)
  470359-688
New Product
 :  MA9016
Milwaukee® pH/ORP Cleaning Solution
  470359-688
New Product
 :  
Milwaukee® pH/ORP Cleaning Solution
Milwaukee® pH/ORP Cleaning Solution
pH/ORP cleaning solution (230 ml)
 :  
$19.25
Each
Premium quality cleaning solution for pH and ORP meters.
  • Factory blended to precise standards in a dedicated, temperature-controlled environment
  • Sealed against light with airtight, tamper proof cap
  • Each bottle marked with lot number and expiration date
Cleaning solution is fundamental for the correct use of electrodes and for obtaining the most accurate and reproducible readings. This solution is formulated to minimize organic growth on your electrode.
Milwaukee’s line of cleaning solutions has been specially formulated to have an expiration of 5 years from the date of manufacture for an unopened bottle. This cleaning solution is designed for use on any pH and ORP probe.
  • Description:
    pH/ORP cleaning solution (230 ml)