Orders LinkContactUsLinkComponent
Solution Bottles, Sterile
Solution Bottles, Sterile
New Product
Solution Bottles, Sterile
New Product
Certificates
Solution Bottles, Sterile
Solution Bottles, Sterile
4 Options Available Below
Product Details & Documents
Solution bottles are key for storage, preparation, and dispensing of chemical solutions and reagents.
  • Compatible with bottle top filters or for use as solution storage containers
    Solution bottle neck size is 45 mm
    Curved bottle design for easy handling
Product Family Options
Product Information
  • Capacity
    Material
    Color
    Availability
    Price
  • 77973-182
    Solution bottle, sterile
    Solution bottle, sterile
    150 ml
    Polystyrene
    Clear
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $100.97
    $100.97
    Case of 24
       $100.97
    Supplier Number: 229781
    Specifications:
    New Product
    New Product
  • 77973-184
    Solution bottle, sterile
    Solution bottle, sterile
    250 ml
    Polystyrene
    Clear
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $109.34
    $109.34
    Case of 24
       $109.34
    Supplier Number: 229782
    Specifications:
    New Product
    New Product
  • 77973-186
    Solution bottle, sterile
    Solution bottle, sterile
    500 ml
    Polystyrene
    Clear
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $150.84
    $150.84
    Case of 24
       $150.84
    Supplier Number: 229784
    Specifications:
    New Product
    New Product
  • 77973-188
    Solution bottle, sterile
    Solution bottle, sterile
    1000 ml
    Polystyrene
    Clear
    Log in for availability
    Log in for availability
    Case of 24
    Case of 24 - $256.26
    $256.26
    Case of 24
       $256.26
    Supplier Number: 229785
    Specifications:
    New Product