Orders LinkContactUsLinkComponent
Milwaukee® Basic Waterproof EC Testers
New Product
Milwaukee® Basic Waterproof EC Testers
New Product
Certificates
Milwaukee® Basic Waterproof EC Testers
4 Options Available Below
Product Details & Documents
These durable EC testers feature a replaceable electrode and are ideal for student use in the classroom and field environments.
  • EC testers that are fast and easy to read with simple one-point calibration
  • Two-year warranty (six months on probe)
  • Automatic temperature compensation (ATC)
  • Over 250 hours of continuous battery life (batteries included)
These models combine a rugged waterproof construction with a replaceable electrode - offering advanced EC accuracy and long term value that outperforms basic testers, making it ideal for repeated use in K-12 science labs and field work.
 : Ordering additional calibration solution is recommended.
 : Batteries, probe, and calibration starter pack included.
Specifications for Product Family
Specifications
  • Catalog No:
  • 470359-568
  • 470359-570
  • 470359-670
  • 470359-668
  • Conductivity Range:
  • 0 - 1999 uS/cm
  • 0.00 - 10.00 mS/cm
  • Conductivity Accuracy:
  • ±2% Full scale
  • ±2% Full scale
Specifications
Product Family Options
EC testers
Product Information
  • Model
    Availability
    Price
  • 470359-568
    Low range EC tester
    Low range EC tester
    C65
    Log in for availability
    Log in for availability
    Each
    Each - $40.65
    $40.65
    Each
       $40.65
    Supplier Number: C65
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    470359-568
    Conductivity Range
    0 - 1999 uS/cm
    Conductivity Accuracy
    ±2% Full scale
    New Product
  • 470359-570
    High range EC tester
    High range EC tester
    C66
    Log in for availability
    Log in for availability
    Each
    Each - $40.65
    $40.65
    Each
       $40.65
    Supplier Number: C66
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    470359-570
    Conductivity Range
    0.00 - 10.00 mS/cm
    Conductivity Accuracy
    ±2% Full scale
    New Product
Replacement probes
Product Information
  • Model
    Availability
    Price
  • 470359-670
    Replacement probe C66
    Replacement probe C66
    Log in for availability
    Log in for availability
    Each
    Each - $32.00
    $32.00
    Each
       $32.00
    Supplier Number: MA73076
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    470359-670
    Conductivity Range
    Conductivity Accuracy
    New Product
  • 470359-668
    Replacement probe for C65
    Replacement probe for C65
    Log in for availability
    Log in for availability
    Each
    Each - $32.00
    $32.00
    Each
       $32.00
    Supplier Number: MA73075
    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    470359-668
    Conductivity Range
    Conductivity Accuracy
    New Product
Calibration solutions
Product Information
  • Catalog #
    Description
    Availability
    Price
  • 1413 µS/cm conductivity solution (230 ml)
    Log in for availability
    Log in for availability
    Each
    Each - $19.25
    $19.25
    Each
       $19.25