Eosin Y modified solution alcoholic solution
Product Details & Documents
Eosin Y solution (modified alcoholic) is a histology and cytology stain offering improved usability for tissue section staining. - Improved usability – lower evaporation rate and enhanced surface tension - Color patterns – stains cytoplasm light pink, collagen pink, muscle pink/rose, and erythrocytes pink/red - Control tissue – suitable for paraffin-embedded, frozen tissue sections, and cell smears
- Constituents: 65% Ethanol, 3.8% Isopropanol, 1% Acetic acid, 0.25% Eosin Y
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Specifications
Specifications
- Size:500 ml