Proteometer-UFT Fluorescent Reagent
Proteometer-UFT Fluorescent Reagent Vials (3X), to be stored at -20 °C
Supplier: NOVILYTIC, LLC
Supplier #: RGN-PUF-001
Proteometer-UFT Fluorescent Reagent
Catalog # 77900-478
Supplier: NOVILYTIC, LLC
New Product
CAS Number:
Jump to:
Product Details & Documents
3 vials of Proteometer-UFT Fluorescent reagent specifically formulated for use with Proteometer-UFT kit.
Vials of the Proteometer-UFT fluorescent reagent for reorder to continue analysis with the Proteometer-UFT kit
Intended for use with the Proteometer-UFT kit.
Ordering information: Reorder in sets of 3 as needed.
Packaging: Supplied in a custom box with instructions for use.
Specifications
Specifications
- Description:Proteometer-UFT Fluorescent Reagent Vials (3X), to be stored at -20 °C