PURELAB® Flex 3+
Product Details & Documents
A unique automated solution providing Type I (ultrapure water) from potable water. Couples directly with analytical chemistry systems delivering complete automation - all in one single unit.
Provides consistent and continuous supply of ultrapure water to analysers to reduce failed runs and loss of samples, reagents and analyser components/accessories
- Provides complete confidence in organic purity and clear display at all times
- The final quality sensor is placed at the entry of the flexible dispenser giving you peace of mind
The compact unit can be placed on the bench or can be wall mounted and has an integrated 7 L reservoir filled by a 20 l/hr. RO membrane, ensuring that water is always available.
Documents
Specifications
- Description:PURELAB® Flex 3+, Type I water quality
- Cat. No.:77918-404
- Flow Rate:2 l/min.
- DNases:<20 pg/ml
- Feed Water:Tap water
- Bacteria:<1 CFU/10 ml
- W×D×H:236×470×1020 mm
- Endotoxin:<0.001 EU/ml
- Toc Content:<5 ppb
- Weight:23 kg
- Inorganics @ 25°C:18.2 MΩ.cm
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



