ContactUsLinkComponent
Capillary Electrophoresis System with UV and Driver for Empower, BioPhase 8800
Capillary Electrophoresis System with UV and Driver for Empower, BioPhase 8800
Catalog # 77999-508
Supplier:  SCIEX
New Product
undefined
Capillary Electrophoresis System with UV and Driver for Empower, BioPhase 8800
Catalog # 77999-508
Supplier:  SCIEX
Supplier Number:  5308400
New Product
Specifications
  • Description:
    BioPhase 8800
Specifications
About this item
BioPhase 8800 capillary electrophoresis system designed for efficiency and ease-of-use operation for purity and heterogeneity analysis of biotherapeutics and other charged molecular species. The BioPhase 8800 system with UV includes integrated UV detection.
  • UV light source and detection optics
  • Touchscreen display allowing selection of pre-developed methods/sequences and system operation (also enables ad hoc system operation)
  • Removable/interchangeable UV filter module housing up to two filters (220 and 280 nm filters included)
The system can be upgraded to include the integrated 488 nm laser (laser sold separately). Includes liquid temperature stabilization module for capillary cartridge temperature control, sample and buffer plate temperature control, integrated touchscreen interface, applications training and a license for the BioPhase 8800 driver for Empower for direct instrument control from waters Empower chromatography data system.
BioPhase 8800 system supply compatibility: BioPhase sample, reagent, and outlet plates designed to ANSI standard configuration; pre-assembled eight-capillary cartridges (bare fused silica or neutral capillary); validated assay kits: BioPhase CE-SDS protein analysis kit, BioPhase Capillary Isoelectric Focusing (cIEF) kit, RNA 9000 purity and integrity kit.