ContactUsLinkComponent
Capillary Electrophoresis PDA System, P/ACE MDQ Plus
Catalog # 77995-572
Supplier:  SCIEX
New Product
undefined
Capillary Electrophoresis PDA System, P/ACE MDQ Plus
Catalog # 77995-572
Supplier:  SCIEX
Supplier Number:  B52521PDA
New Product
Specifications
  • Description:
    P/ACE MDQ Plus
Specifications
About this item
Automated programmable methods and sample handling capabilities for walk-away operation. Programmable multimode sample introduction; electrokinetic, pressure or vacuum.
  • Programmable internal fluidic system generates pressure or vacuum from −5 to +100 psi
  • Sample storage temperature control
  • Exclusive circulating liquid cooling system that precisely regulates capillary temperature
  • Direct control provides intuitive management of all system functions in real time
  • Starter kit containing necessary hardware supplies to operate the system
  • Installation Qualification (IQ1) and Operation Qualification (OQ1) of the system
  • System installation and training
The P/ACE MDQ Plus includes High-resolution separation module; Modular Diode Array Detector, Interface Board and Optics cable; High-speed system controller and with 32 Karat™ Software Version 10.3 running in a Windows 10 environment providing instrument control, data acquisition and analysis; 22” Flat Panel Display; and Built in auto-sampler and programmable array of 36 buffer pairs.