Specifications
- Description:BioPhase 8800
Specifications
About this item
BioPhase 8800 capillary electrophoresis system designed for efficiency and ease-of-use operation for purity and heterogeneity analysis of biotherapeutics and other charged molecular species. The BioPhase 8800 system with UV includes integrated UV and laser-induced fluorescence (488 nm laser) detection.
UV light source and detection optics
- Integrated 488 nm laser source and detection (source/detection system is self-calibrating and are controlled via software)
- Touchscreen display allowing selection of pre-developed methods/sequences and system operation (also enables ad hoc system operation)
- Integrated networking capability allowing for system operation via SCIEX control software enabling method development, sequence creation, and data analysis
- Removable/interchangeable UV filter module housing up to 2 filters (220 and 280 nm filters included)
- Interchangeable LIF filter capability
Includes liquid temperature stabilization module for capillary cartridge temperature control, sample and buffer plate temperature control, integrated touchscreen interface, applications training and a license for the BioPhase 8800 driver for Empower for direct instrument control from waters empower chromatography data system.
BioPhase 8800 system supply compatibility: BioPhase sample, reagent, and outlet plates designed to ANSI standard configuration; Pre-assembled 8-capillary cartridges (bare fused silica or neutral capillary); validated assay kits: BioPhase CE-SDS protein analysis kit, BioPhase Capillary Isoelectric Focusing (cIEF) kit, RNA 9000 purity and integrity kit.