Order Entry
United States
ContactUsLinkComponent
Human Recombinant G-CSF (from E. coli)
Human Recombinant G-CSF (from E. coli)
Supplier:  Sartorius
New Product
Certificates
undefined
Human Recombinant G-CSF (from E. coli)
Supplier:  Sartorius
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    Ambient
  • Endotoxin-free:
    Y
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    Granulocyte Colony-Stimulating Factor
  • Protein/Peptide Name:
    G-CSF
  • Nuclease-free:
    N
  • Shipping Temperature:
    Ambient

Specifications

Related Information

Frequently Bought Together