- Description:3Doodler FLOW essentials pen set
Versatile 3D printing pen with LCD display for creative projects.
Compatible with 1.75 mm PLA and ABS filaments
Integrated LCD for real-time temperature monitoring
Dual-drive technology for improved control and ergonomics
Designed for creators aged 14 and up
The 3Doodler FLOW Essentials 3D Printing Pen Set offers a hands-on approach to 3D creation, allowing users to draw and build three-dimensional objects with ease.
Its dual-drive technology ensures smooth filament extrusion, while the integrated LCD provides precise temperature control for various filament types. This pen is ideal for artists, hobbyists, and educators seeking an interactive tool to bring ideas to life.
Kit Includes: (1) 3Doodler Flow 3D printing pen, (8) PLA filament spools (1.75 mm) in assorted colors, (1) stencil sheet, (1) ultimate guide to doodling, (1) power adapter, (1) maintenance tool.