Human Recombinant M-CSF (from E. coli)
Supplier: Sartorius
New Product
Certificates
About this item
Manufactured to the highest quality standards using recombinant DNA technology which do not contain any animal-derived components or contaminants.
Specifications
- Conjugation:Unconjugated
- Protein Function:Cytokine
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Storage Conditions:Ambient
- Endotoxin-free:Y
- Carrier-Free:Y
- Protease-free:N
- Animal-Free:Y
- Protein Synonyms:Macrophage-Colony Stimulating Factor
- Protein/Peptide Name:M-CSF
- Nuclease-free:N
- Shipping Temperature:Ambient
Specifications
Frequently Bought Together