NEBNext® Multiplex Oligos for Illumina® (Dual Index Primers Set 1)
Catalog # 102877-580
Supplier: New England Biolabs (NEB)
Small Business Enterprise
CAS Number:
Specifications
- Description:NEBNext® Multiplex oligos for Illumina® (dual index primers set 1)
- Cat. No.:102877-580
- No. of reactions:96
- Supplier no.:E7600S
Specifications
About this item
NEBNext Multiplex oligos provide adaptors and primers to enable high yield multiplex Illumina library production.
High efficiency adaptor ligation
- Minimized adaptor-dimer formation
- Includes index primers for library multiplexing
- Increased sample identification specificity
- Includes USER Enzyme
The unique hairpin loop structure of the NEBNext adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. This kit includes 8 i5 index primers and 12 i7 index primers for dual indexing. Additional NEBNext Multiplex Oligos products are available for different indexing strategies.