NEBNext® Multiplex Oligos for Illumina® (Index Primers Set 2)
Supplier: New England Biolabs (NEB)
Certificates
About this item
NEBNext Multiplex oligos provide adaptors and primers to enable high yield multiplex Illumina library production.
High efficiency adaptor ligation
- Minimized adaptor-dimer formationIncludes index primers for library multiplexing
- Single index primers for multiplexing
- Includes USER Enzyme
The unique hairpin loop structure of the NEBNext adaptor minimizes adaptor-dimer formation, and NEBNext index PCR primers enable index incorporation during library amplification. Additional NEBNext multiplex oligos products are available for different indexing strategies.