About this item
Accelerate research with the Eppendorf™ CellXpert™ CS220 CO₂ Incubator Shaker, the only CO₂ incubator shaker with integrated 180 °C sterilization. Achieve high flask capacity and reliable results with optimized lab space efficiency and low operating costs.
High flask capacity
- 180 °C sterilization routine integrated: including wizard and downloadable performance protocol, no locally restricted UV-airflow/surface-disinfection
- Stainless steel chamber: hygienic design according to EN ISO 14159:2008 standard plus only few internal parts and no drive wheel, crevices, cables, or heating fins inside
- Optimized throughput per lab space: highest platform size vs. footprint ratio in the market, stackable up to three-fold
- Saves costs: includes slide-out platform, rH-control, touch interface, access port, 180 °C sterilization routine, no UV-lamps that require frequent exchange, low gas and energy consumption
- Eppendorf X-drive: proven and tested shaker drive with four weight-supporting shafts for 24/7 vibration-free shaking
- Glide-up door and slide-out platform: saves space and ensures fast and easy experiment setup, cleaning, and service (even at the back of the chamber)
- Full control and fast root-cause analysis: 7" touchscreen shows all setpoints and current parameters at a glance, direct visual overview of logged data/events including door openings and alarms, user management
The CellXpert™ CS220 is a large capacity, stackable CO₂ incubator shaker that was designed to accelerate mammalian suspension culture research projects. Applications include protein expression, screening, cell line development, early process development, viral vector production in mammalian cells (e.g., Ad, AAV, LV), bioreactor inoculum preparation.
Please order separately: platform, clamps, sticky pads, storage base or feet, water supply package. Optional: 1 to 2 stacking kits and water tank holder.
Specifications
- Catalog No:
- 78000-726
- 78000-694
- 78000-734
- 78000-718
- 78000-720
- 78000-722
- 78000-724
- 78000-732
- 78000-714
- 78000-716
- Temperature Stability:
- 0.1 °C
- 0.1 °C
- Temperature Uniformity:
- ±0.3 °C at 37 °C
- ±0.3 °C at 37 °C
- Temperature Accuracy:
- ±0.4 °C at 37 °C
- ±0.4 °C at 37 °C
- Permissible Relative Humidity:
- Up to 85%
- Up to 85%
- Drive Type:
- Eppendorf X-drive
- Eppendorf X-drive
- Display:
- Color Touch Screen
- Color Touch Screen