About this item
The LHS and LHS Flow are compact benchtop automated liquid handlers with intuitive method development software, an outstanding solution for automating common repetitive pipetting tasks. The LHS easily prepares ELISA, PCR, serial dilutions, cherry picking, aliquoting and more. Units provide all the benefits from automation with none of the complexity.
Automates pipetting tasks, allowing more time for other work
- Quiet operation with adjustable settings for various workflows
- Cost-effective automation for small to midsize sample volumes
- Volume range for 1-channel: 1 to 50 µl, 10 to 200 µl, 40 to 1000 µl
- Volume range for 8-channel: 1 to 50 µl, 20 to 300 µl, 40 to 1000 µl
- Working positions: 8 positions (P2-P8); P1 for waste box
- Applications: PCR/qPCR, ELISA, enzyme assay, serial dilutions, cherry picking, plate replication and more
The Liquid Handling Station (LHS) and Liquid Handling Station Flow (LHS Flow) streamline pipetting workflows with user-friendly software, quiet operation, and adaptable settings. BRANDTECH's highly trained team is available to support user needs to ensure success, with a focus on ensuring performance by integrating the LHS into existing workflows. BRANDTECH and VWR provide users with support at every step along the way, from the selection process, to order placement, and of course, during delivery, set-up, and training.
Intuitive: create methods in minutes using LHS’s intuitive software interface, with no programming needed.
Versatile: seven configurable work positions accommodate individual microplates (24,96,384), tubes, vials, and any other vessels required to run assays/tests.
Compact: with a footprint of 24×20" and 21" tall, the LHS fits right on the lab bench, with one of the smallest footprints in the industry. Units fit easily under overhead cabinets, freeing up valuable overhead storage.
Consistent: eliminates pipetting errors from monotonous, repetitive tasks, producing consistent results and saving both time and budget.
For applications requiring a controlled environment, the Liquid Handling Station Flow adds a built-in horizontal laminar air flow box to ensure contamination-free liquid handling. Meets ISO 14664-1 (Class 5) and GMP Annex 1 (Class A) standards.
Designed for efficiency and reliability, both systems offer flexible configurations for various lab applications.
Both units include: motor control unit, control software, operating manual, power cable, USB cable, documentation, and on-site installation and training. The Liquid Handling System Flow also includes: Flow box filtration unit and USB hub.
Specifications
- Catalog No:
- 77775-520
- 77775-522
- Model:
- Liquid handling station
- Liquid handling station flow
- Power:
- 100 - 240 V, 50/60 Hz
- 100 - 240 V, 50/60 Hz
- W×D×H of system:
- Door open: 595×485×690 mm (23.43×19.09×27.17") Door closed: 595×485×530 mm (23.43×19.09×20.87")
- Door open: 595×515×662 mm (23.43×20.28×26.06") Door closed: 595×515×662 mm (23.43×20.28×26.06")
- Weight Of System (kg):
- approx. 25 kg (55.1 lb)
- approx. 35 kg (77.2 lb)