Transfer Caps for VWR® Erlenmeyer Shaker Flasks
Catalog # 77889-502
Supplier: VWR International
New Product
Specifications
- Sterility:Sterile
- Description:Transfer cap of 5 L Erlenmeyer flasks, with Male MPC tubing connection
- Length:Dip tube length: 33.5 cm, Int. Ø: 3.9 mm