United Scientific Equal Volume Density Cubes
470356-008
- Description:Equal volume density cubes
Economical classroom solution for density lessons.
Uniform in design
- Variety of materials
- 0.5" cubes
The set consists of five cubes, all of equal volume. Each sides measures 0.5", and come in five different materials. The cubes are aluminum, steel, acrylic, polypropylene, and PVC. The variety of materials allows users to investigate materials that float (PP) and sink.