Order Entry
United States
ContactUsLinkComponent
Octylphenolpoly(ethyleneglycolether)x (Triton X-100 Substitute), 2% for Biochemistry Class
Octylphenolpoly(ethyleneglycolether)x (Triton X-100 Substitute), 2% for Biochemistry Class
Catalog # 76286-404
Supplier:  G-Biosciences
Octylphenolpoly(ethyleneglycolether)x (Triton X-100 Substitute), 2% for Biochemistry Class
Catalog # 76286-404
Supplier:  G-Biosciences
Supplier Number:  BTNM-0077

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Octylphenolpoly(ethyleneglycolether)x (Triton® X-100 substitute), 2%
  • Size:
    20 ml
  • Grade:
    Research
  • Form:
    2% Aqueous clear solution (v/v)
  • Type:
    Non-ionic detergent

Specifications

About this item

Immobilized Octylphenolpoly(ethyleneglycolether)x is designed to allow refolding of recombinant proteins that were aggregated or formed inclusion bodies when expressed in the genetically engineered cells.

  • Part of Ellyn Daugherty's Biotechnology: Science for the New Millennium

Biotechnology: Science for the New Millennium allows students to increase conceptual background as they build lab proficiencies. Science-bound and business-bound students benefit from the extensive skill development, science methods, and career exploration that permeate the curriculum.