Kidder Wooden House Frame STEM Build Kit
470358-038
New Product
- Description:Wooden house frame STEM build kit
Assemble a wooden house frame to explore architectural design and structural principles.
- Encourages hands-on STEM learning Demonstrates principles of triangle stability and rectangular balance No cutting required; glue included Includes online teacher resource guide
The Wooden House Frame STEM Build Kit offers students an engaging hands-on experience to explore architectural design and structural engineering principles. By assembling a model of a wooden house frame, learners gain insight into how triangle stability and rectangular balance contribute to the integrity of structures. This activity not only reinforces theoretical knowledge but also enhances problem-solving skills, making it an excellent addition to STEM education programs.
Application: Engineering, Mechanics