Kidder Hydraulic Excavator STEM Build Kit
Catalog # 470358-040
Supplier: KIDDER MFG. CO. LTD
New Product
Specifications
- Description:Hydraulic excavator STEM build kit
Specifications
About this item
Build a working excavator to explore hydraulic mechanics in a fun, hands-on way.
- Demonstrates hydraulic principles through assembly
- Encourages hands-on STEM learning
- Simple setup suitable for classroom use
- Affordable and compact design
The Hydraulic Excavator STEM Build kit offers students an engaging introduction to hydraulic systems and mechanical engineering principles. By assembling the excavator, learners explore how fluid pressure can be used to create mechanical functions, providing a tangible demonstration of hydraulic energy concepts. This hands-on activity not only reinforces theoretical knowledge but also enhances problem-solving skills.
Application: Engineering, Mechanics
Delivery: Kit includes Pre-cut wooden parts, syringes and tubing for hydraulic system, assembly instructions.