Orders LinkContactUsLinkComponent
Basic Latent Print Kits
Basic Latent Print Kits
New Product
Basic Latent Print Kits
New Product
Certificates
Basic Latent Print Kits
Basic Latent Print Kits
2 Options Available Below
Product Details & Documents
Great for classroom use.
  • Full set of materials in a single pack
  • Standard black or high visibility orange storage box
Use for lifting latent fingerprints from standard non-porous surfaces like glass, and smooth painted surfaces. Each kit includes a case with a foam liner to keep backers, tape, brush, and powder separated.
Product Family Options
Product Information
  • Availability
    Price
  • 470358-948
    Standard fingerprint kit, orange case
    Standard fingerprint kit, orange case
    Log in for availability
    Log in for availability
    Each
    Each - $70.55
    $70.55
    Each
       $70.55
    Supplier Number: 40023
    Specifications:
    New Product
    New Product
  • 470358-910
    Standard fingerprint kit, black case
    Standard fingerprint kit, black case
    Log in for availability
    Log in for availability
    Kit of 1
    Kit of 1 - $74.60
    $74.60
    Kit of 1
       $74.60
    Supplier Number: 05600
    Specifications:
    New Product
    New Product