Specifications
- Description:Spin touch
Specifications
About this item
Effortlessly test fresh, salt, and brackish water in just 2 minutes with the most advanced, all-in-one photometer designed for fish farming applications.
Tests fresh, salt, and brackish water with one device
- Delivers accurate results in just 2 minutes
- Uses pre-filled reagent disks - no measuring or mixing
- Eliminates vials, prep time, and clean-up
- Simple operation - anyone can use it
- Ideal for fish farming and aquaculture applications
- Compact and portable for easy field testing
The WaterLink® Spin Touch® FX is a user-friendly, all-in-one water testing tool designed for teachers to engage students in hands-on environmental science. Using sealed SpinDisk® reagent cartridges, students can easily test fresh, salt, or brackish water samples and get accurate results in just 2 minutes. This compact, portable photometer eliminates the need for vials or complex prep, allowing teachers to focus on instruction while students gain practical experience. Ideal for biology, environmental science, or marine science lessons, it’s the perfect tool to bring real-world water testing into the classroom or field.
Kit includes water sample bottle (60 ml), plastic beaker, 50 ml, syringe with tips (3), syringe tips (3), cleaning tissues, meter check disk, universal disk cover, USB cable, AC adapter (North America), cloth wipe, mobile bluetooth printer.