ContactUsLinkComponent
Total Bacteria Slides
  470358-622
New Product
undefined
Total Bacteria Slides
  470358-622
 :  CM858MC
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Total bacteria slides
  • Environmentally Preferable:
  • No. of slides:
    5
Used to monitor microbial contamination.
  • Tests for the pressence of bacteria
  • Also known as a dip slide
  • No special equipment or training necessary
  • MS/HS grade level activity
Supplied with test instructions and results chart. Results in 24 to 48 hours after incubation. Resulting growth patterns are compared to the included chart to ascertain the level of microbial contamination.