Duo Biopaddle Slides
470358-624
New Product
- Description:Duo biopaddles slides
- No. of users:5
Used for monitoring microbial activity in liquid.
Can test for both fungi and bacteria
- Also known as a dip slide
- No special equipment or training necessary
- MS/HS grade level activity
Supplied with test instructions and comparison chart. Easily interpreted test results in 24 to 48 hours for bacteria growth, and three to five days for fungal growth.