ContactUsLinkComponent
Kidde Pro-5MP Fire Extinguisher, with Wall Hanger, 3-A, 40-B:C
Kidde Pro-5MP Fire Extinguisher, with Wall Hanger, 3-A, 40-B:C
Catalog # 77929-484
New Product
Kidde Pro-5MP Fire Extinguisher, with Wall Hanger, 3-A, 40-B:C
Catalog # 77929-484
Supplier Number:  KID21032142
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Pro-5MP series fire extinguisher, with wall hanger
Specifications
About this item
Stop common fires involving trash, wood, paper, liquids, gases, and electrical equipment.
  • All-metal construction with metal valve assembly
  • 3× more firefighting power
  • Rechargeable
  • UL rating: 3-A, 40-B:C
  • Weight: 5.5 lbs.
Safeguard any property with the dependable Kidde PRO-5MP rechargeable fire extinguisher, designed for commercial use. This extinguisher is designed to meet code compliance for larger areas and common fires involving wood, paper, and textiles (Class A), flammable liquids and gases (Class B), and electrical equipment (Class C). Manufactured from durable, corrosion-resistant aluminum, the Kidde Pro fire extinguisher is a breeze to handle. It provides a remarkable discharge rate of 13 to 15 seconds, a range of 12 to 18 feet, and an operating pressure of 195psi.
The sleek, painted aluminum handle, aluminum cylinder, and metal valve are impact-resistant, making it dependable when needed most. The easy-to-pull safety pin provides quick access for simple operation designed to combat fire hazards using the PASS (Pull, Aim, Squeeze, Sweep) firefighting method. The multi-directional discharge hose allows action from various angles. Note that the dry chemical agent in this fire extinguisher is non-toxic but may cause skin irritation. Place a fire extinguisher within easy reach where fires are more likely to start. Rechargeable.