Specifications
- Description:Energizer® industrial lithium CR2 photo batteries, 3V
Specifications
About this item
Rely on energizer industrial lithium CR2 batteries to power specialty professional devices.
Keep all specialty devices powered up for work
- Safeguard professional equipment with lithium batteries that are 100% leakproof in standard usage
- Get remarkable performance, even in extreme work site temperatures
Trust your precision electronics to lithium CR2 batteries that are 100% leakproof in normal usage. They operate in work site temperatures ranging from −40 to +140 °F. They hold their power with up to ten years of shelf life, virtually eliminating device downtime. When work depends on laser pointers, motion sensors, automatic paper towel dispensers, and more, rely on the power and protection of energizer industrial lithium CR2 batteries.