ContactUsLinkComponent
Innovating Science® Antimicrobial Properties of Essential Oils Lab Kit
Innovating Science® Antimicrobial Properties of Essential Oils Lab Kit
Catalog # 470357-888
Supplier:  Ward's Science
New Product
undefined
Innovating Science® Antimicrobial Properties of Essential Oils Lab Kit
Catalog # 470357-888
Supplier:  Ward's Science
Supplier Number:  IS3055
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Antimicrobial properties of essential oils lab kit
  • No. of lab groups:
    15
Specifications
About this item
Explore the antimicrobial efficacy of essential oils commonly used in household cleaning products.
  • Kit contains enough materials for 15 groups Teacher's manual and student study guide copymaster included
Essential oils have been used for a variety of purposes throughout history for their scents, flavors, and pharmacological properties. Some oils are commonly used in cleaning products, but which ones are most effective? In this activity, students will perform the Kirby-Bauer test on a variety of essential oils to determine their antimicrobial efficacy.
Ordering information: Required but not included: Test tubes, test tube racks, tape, markers, metric rulers, hot water bath, biohazard waste receptacle.
Delivery information: Kit includes: Peppermint oil, 10 ml; Lavender oil, 10 ml; Cinnamon oil, 10 ml; Lemon oil, 10 ml; Balsam oil, 10 ml; 2× nutrient agar, 200 ml; 3× nutrient broth tubes, 9 ml; 15× Sterile cotton swabs, 2-pack; 20× petri dishes; 2× blank sterile disks, 50-pack; 5× forceps.