ContactUsLinkComponent
Innovating Science® Synthesis of a Coordination Complex Lab Kit
Innovating Science® Synthesis of a Coordination Complex Lab Kit
Catalog # 470357-890
Supplier:  Ward's Science
New Product
undefined
Innovating Science® Synthesis of a Coordination Complex Lab Kit
Catalog # 470357-890
Supplier:  Ward's Science
Supplier Number:  IS2535
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Synthesis of a coordination complex lab kit
  • No. of lab groups:
    15
Specifications
About this item
Produce a bright-green iron compound in this multi-step synthesis.
  • Kit contains enough materials for 15 groups Teacher's manual and student study guide copymaster included
Coordination complexes are brightly colored compounds consisting of a central metal atom and ligand molecules, atoms, or ions. Synthesize and purify a lime-green coordination complex called potassium ferrioxalate trihydrate in this multi-step synthesis and optionally analzye the complex via UV-Visible spectrophotometry.
Ordering information: Required but not included: 150 ml Glass beakers, funnels, hot plate stirrers, magnetic stir bars, thermometers, distilled or deionized water, laboratory scale, 25 or 50 ml graduated cylinders, tongs or heat-resistant gloves, forceps, spatulas, ice bath, 500 ml glass beakers, cuvettes [optional], UV-Visible spectrophotometer [optional].
Delivery: Kit includes: Oxalic acid 0.5 M solution, 800 ml; Potassium oxalate 1.0 M solution, 225 ml; Hydrogen peroxide 3% solution, 225 ml; Ethanol, 95%, 150 ml; Ethanol, 50%, 225 ml; Sulfuric acid, 0.1 M, 25 ml; Ferrous ammonium sulfate hexahydrate, 50 g; 30× pipettes; 15× filter papers.