ContactUsLinkComponent
Innovating Science® Introduction to Complexometric Titration Lab Kit
Innovating Science® Introduction to Complexometric Titration Lab Kit
Catalog # 470357-876
Supplier:  Ward's Science
New Product
undefined
Innovating Science® Introduction to Complexometric Titration Lab Kit
Catalog # 470357-876
Supplier:  Ward's Science
Supplier Number:  IS2534
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Introduction to complexometric titration lab kit
  • No. of lab groups:
    15
Specifications
About this item
Perform a common analytical technique, complexometric titration, to quantify calcium in samples.
  • Kit contains enough materials for 15 groups Teacher's manual and student study guide copymaster included
Complex formation is frequently used in both qualitative and quantitative inorganic analysis. Introduce topics such as metal-ligand interactions in this activity using complexometric titration to determine the calcium content of both known and unknown samples.
Ordering information: Required but not included: 125 ml Erlenmeyer flasks, 50 ml burettes, ring stands with burette clamps, 10 or 25 ml graduated cylinders, funnels, distilled or deionized water, white paper [optional], magnetic stir plates and stir bars [optional].
Delivery: Kit includes: 4× EDTA 0.1 M solution, 500 ml; 3× Eriochrome black T (EBT) indicator solution, 60 ml; Calcium chloride 0.1 M solution, 500 ml; unknown Calcium solution #1, 500 ml; unknown Calcium solution #2, 500 ml; 3× pH 10 buffer solution 500 ml.