ContactUsLinkComponent
Innovating Science® Firefly Oscillating Chemiluminescence Demo
Catalog # 470357-516
Supplier:  Ward's Science
New Product
undefined
Innovating Science® Firefly Oscillating Chemiluminescence Demo
Catalog # 470357-516
Supplier:  Ward's Science
Supplier Number:  IS7052
New Product
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Firefly oscillating chemiluminescence demo
Specifications
About this item
A glow-in-the-dark chemical reaction resembling a firefly in the summer.
  • Includes an instruction manual with background information
  • Enough materials for three demonstrations
Two glow-in-the-dark reactions occur in turn: one that is bright and quick, and another that is dull and slow. The oscillation between these reactions looks like a firefly on a summer night! There are enough materials to perform this demonstration three times.
Ordering information:
Required but not included: 2 L Beaker, magnetic stir bar, hot plate stirrer, thermometer.
Delivery:
Kit includes: 3× Luminol-Thiocyanate solution, 900 ml; 3× Copper sulfate solution, 100 ml; 3× Hydrogen peroxide, 30%, 30 ml.