

Acetonitrile ≥99.9% tested for EPA533 and EPA537.1 PFAS Methods, LiChrosolv®, Supelco®
Catalog # EM1047261000
Supplier: MilliporeSigma
New Product
CAS Number:
Specifications
- Pack type:Borosilicate glass bottle
- Size:1 L
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental Certification:Chem21 solvent selection guide recommended by default
Specifications
Specification Test Results
- EPA 533/537.1 Tested:Conforms
- UHPLC Gradient (at 210 nm):≤1.0 mAU
- Evaporation Residue:≤ 0.7 mg/l
- Titratable Acid (meq/g):≤ 0.001
- Titratable Base (meq/g):≤ 0.0001
Specification Test Results
About this item
CAS Number:75-05-8
:
Formula:C₂H₃N
MDL Number:MFCD00001878
:41.05 g/mol
UN:1648
REACH:01-2119471307-38-XXXX
:GHS02
:GHS07
Storage Temperature:Room Temperature